Search
Search

SLC6A9| Gene Symbol: SLC6A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005607116.3 | XP_005607173.1 | 1839 | 66 | ||
| XM_014737476.2 | XP_014592962.1 | 1683 | 66 | |||
| XM_014737477.2 | XP_014592963.2 | 1747 | 66 | |||
| XM_023631525.1 | XP_023487293.1 | 1802 | 66 | |||
| XM_023631531.1 | XP_023487299.1 | 1517 | 66 | |||
| XM_023631535.1 | XP_023487303.1 | 1600 | 66 | |||
| XM_023631536.1 | XP_023487304.1 | 1886 | 66 | |||
| XM_023631539.1 | XP_023487307.1 | 1513 | 66 | |||
Set Membership: |
|