Search
Search
| Gene Symbol: RERE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014737639.2 | XP_014593125.2 | 2262 | 81 | ||
| XM_023635850.1 | XP_023491618.1 | 1277 | 81 | |||
| XM_023635851.1 | XP_023491619.1 | 1141 | 81 | |||
| XM_023635852.1 | XP_023491620.1 | 2271 | 81 | |||
| XM_023635853.1 | XP_023491621.1 | 2136 | 81 | |||