Search
Search

ACAP3| Gene Symbol: ACAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023636031.1 | XP_023491799.1 | 493 | 59 | ||
| XM_023636033.1 | XP_023491801.1 | 382 | 59 | |||
| XM_023636034.1 | XP_023491802.1 | 382 | 59 | |||
Set Membership: |
|