Search
Search

COL9A1| Gene Symbol: COL9A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023625140.1 | XP_023480908.1 | 2053 | 90 | ||
| XM_023625141.1 | XP_023480909.1 | 2050 | 90 | |||
| XM_023625142.1 | XP_023480910.1 | 867 | 90 | |||
| XM_023625143.1 | XP_023480911.1 | 919 | 90 | |||
Set Membership: |
|