Search
Search

SLC17A9| Gene Symbol: SLC17A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005604852.3 | XP_005604909.1 | 728 | 98 | ||
| XM_023626897.1 | XP_023482665.1 | 743 | 98 | |||
| XM_023626898.1 | XP_023482666.1 | 729 | 98 | |||
| XM_023626900.1 | XP_023482668.1 | 631 | 98 | |||
| XM_023626901.1 | XP_023482669.1 | 729 | 98 | |||
| XM_023626902.1 | XP_023482670.1 | 626 | 98 | |||
Set Membership: |
|