Search
Search
| Gene Symbol: DLC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005606354.3 | XP_005606411.1 | 3345 | 56 | ||
| XM_014736595.2 | XP_014592081.1 | 3108 | 56 | |||
| XM_023630733.1 | XP_023486501.1 | 3220 | 56 | |||
Set Membership: |
|