Search
Search

ATP2B1| Gene Symbol: ATP2B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023631606.1 | XP_023487374.1 | 2441 | 65 | ||
| XM_023631607.1 | XP_023487375.1 | 2441 | 65 | |||
| XM_023631608.1 | XP_023487376.1 | 2441 | 65 | |||
Set Membership: |
|