Search
Search

NCAPH2| Gene Symbol: NCAPH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014736960.2 | XP_014592446.1 | 1563 | 80 | ||
| XM_014736961.2 | XP_014592447.1 | 1560 | 80 | |||
| XM_014736962.2 | XP_014592448.1 | 1561 | 80 | |||
| XM_014736963.2 | XP_014592449.1 | 1558 | 80 | |||