Search
Search

HSD17B11| Gene Symbol: HSD17B11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001495980.4 | XP_001496030.1 | 428 | 100 | ||
| GenBank mRNA | HQ890019.1 | - | 2 - 3 | 364 | 100 | |
| JL616172.2 | - | 2 - 3 | 479 | 100 | ||
Set Membership: |
|