Search
Search

PSEN2| Gene Symbol: PSEN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005607965.3 | XP_005608022.2 | 575 | 90 | ||
| XM_005607966.3 | XP_005608023.2 | 577 | 90 | |||
| XM_005607967.3 | XP_005608024.2 | 579 | 90 | |||
| XM_023632694.1 | XP_023488462.1 | 580 | 90 | |||
| XM_023632695.1 | XP_023488463.1 | 418 | 90 | |||
| XM_023632697.1 | XP_023488465.1 | 835 | 90 | |||
| XM_023632698.1 | XP_023488466.1 | 687 | 90 | |||
| XM_023632699.1 | XP_023488467.1 | 666 | 90 | |||
| XM_023632700.1 | XP_023488468.1 | 575 | 90 | |||
| XM_023632701.1 | XP_023488469.1 | 545 | 90 | |||
| XM_023632702.1 | XP_023488470.1 | 581 | 90 | |||
| XM_023632704.1 | XP_023488472.1 | 582 | 90 | |||
Set Membership: |
|