Search
Search

SEC14L2| Gene Symbol: SEC14L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001498151.5 | XP_001498201.1 | 1046 | 87 | ||
| XM_005612435.3 | XP_005612492.1 | 1210 | 87 | |||
| XM_005612436.3 | XP_005612493.1 | 955 | 87 | |||
| XM_023646856.1 | XP_023502624.1 | 1136 | 87 | |||