Search
Search
| Gene Symbol: NF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001498860.6 | XP_001498910.1 | 1353 | 134 | ||
| XM_005612454.3 | XP_005612511.1 | 1354 | 134 | |||
| XM_023646882.1 | XP_023502650.1 | 1354 | 134 | |||
| XM_023646883.1 | XP_023502651.1 | 1354 | 134 | |||
Set Membership: |
|