Search
Search

GREB1L| Gene Symbol: GREB1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023647512.1 | XP_023503280.1 | 6059 | 131 | ||
| XM_023647513.1 | XP_023503281.1 | 19978 | 131 | |||
| XM_023647514.1 | XP_023503282.1 | 6148 | 131 | |||
| XM_023647515.1 | XP_023503283.1 | 6256 | 131 | |||
| XM_023647517.1 | XP_023503285.1 | 5890 | 131 | |||
| XM_023647518.1 | XP_023503286.1 | 5869 | 131 | |||
| XM_023647519.1 | XP_023503287.1 | 5952 | 131 | |||
| XM_023647520.1 | XP_023503288.1 | 6055 | 131 | |||
| XM_023647521.1 | XP_023503289.1 | 5815 | 131 | |||
Set Membership: |
|