Search
Search

UBXN2B| Gene Symbol: UBXN2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005614886.3 | XP_005614943.2 | 725 | 109 | ||
| XM_023648562.1 | XP_023504330.1 | 798 | 109 | |||
| XM_023648563.1 | XP_023504331.1 | 938 | 109 | |||
| XM_023648564.1 | XP_023504332.1 | 873 | 109 | |||
Set Membership: |
|