Search
Search

CIART| Gene Symbol: CIART | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014739786.2 | XP_014595272.1 | 684 | 89 | ||
| XM_014739790.2 | XP_014595276.1 | 428 | 89 | |||
| XM_014739791.2 | XP_014595277.1 | 526 | 89 | |||
| XM_014739792.2 | XP_014595278.1 | 362 | 89 | |||
| GenBank mRNA | JL618614.1 | - | 3 - 4 | 250 | 89 | |
Set Membership: |
|