Search
Search

FBXL4| Gene Symbol: FBXL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001503857.6 | XP_001503907.1 | 1981 | 70 | ||
| XM_005596840.3 | XP_005596897.1 | 1952 | 70 | |||
| XM_014734214.1 | XP_014589700.1 | 1911 | 70 | |||
| XM_014734220.2 | XP_014589706.1 | 1911 | 70 | |||
| XM_023650840.1 | XP_023506608.1 | 1793 | 70 | |||
Set Membership: |
|