Search
Search

CEP131| Gene Symbol: CEP131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023651815.1 | XP_023507583.1 | 1718 | 93 | ||
| XM_023651816.1 | XP_023507584.1 | 1716 | 93 | |||
| XM_023651818.1 | XP_023507586.1 | 1719 | 93 | |||
| XM_023651819.1 | XP_023507587.1 | 1718 | 93 | |||
| XM_023651820.1 | XP_023507588.1 | 1715 | 93 | |||
| XM_023651821.1 | XP_023507589.1 | 1719 | 93 | |||