Search
Search

LUC7L3| Gene Symbol: LUC7L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023652757.1 | XP_023508525.1 | 496 | 123 | ||
| XM_023652758.1 | XP_023508526.1 | 496 | 123 | |||
| XM_023652759.1 | XP_023508527.1 | 496 | 123 | |||
| XM_023652760.1 | XP_023508528.1 | 495 | 123 | |||
| XM_023652761.1 | XP_023508529.1 | 495 | 123 | |||
| XM_023652762.1 | XP_023508530.1 | 496 | 123 | |||
| XM_023652763.1 | XP_023508531.1 | 579 | 123 | |||
| XM_023652764.1 | XP_023508532.1 | 263 | 123 | |||
| GenBank mRNA | JL616221.1 | - | 2 - 3 | 191 | 123 | |
Set Membership: |
|