Search
Search

PELP1| Gene Symbol: PELP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001502967.3 | XP_001503017.3 | 836 | 102 | ||
| XM_023653374.1 | XP_023509142.1 | 645 | 102 | |||
| XM_023653375.1 | XP_023509143.1 | 651 | 102 | |||
Set Membership: |
|