Search
Search

CTNND1| Gene Symbol: CTNND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001497191.6 | XP_001497241.5 | 2861 | 69 | ||
| XM_005598153.3 | XP_005598210.2 | 2879 | 69 | |||
| XM_005598154.3 | XP_005598211.2 | 2880 | 69 | |||
| XM_005598155.3 | XP_005598212.2 | 2862 | 69 | |||
| XM_005598156.3 | XP_005598213.2 | 2879 | 69 | |||
| XM_005598158.3 | XP_005598215.2 | 2473 | 69 | |||
| XM_014729523.2 | XP_014585009.2 | 2763 | 69 | |||
| XM_023654081.1 | XP_023509849.1 | 2806 | 69 | |||
| XM_023654082.1 | XP_023509850.1 | 2880 | 69 | |||
Set Membership: |
|