Search
Search
| Gene Symbol: CTH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005610499.3 | XP_005610556.1 | 1210 | 58 | ||
| XM_023641855.1 | XP_023497623.1 | 1195 | 58 | |||
| GenBank mRNA | JL637288.1 | - | 5 - 6 | 470 | 58 | |
Set Membership: |
|