Search
Search

ATG2A| Gene Symbol: ATG2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014729681.2 | XP_014585167.1 | 2209 | 114 | ||
| XM_023654382.1 | XP_023510150.1 | 2212 | 114 | |||
| XM_023654383.1 | XP_023510151.1 | 2209 | 114 | |||
| XM_023654384.1 | XP_023510152.1 | 2212 | 114 | |||
Set Membership: |
|