Search
Search

LOC100067674| Gene Symbol: LOC100067674 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023617042.1 | XP_023472810.1 | 779 | 75 | ||
| XM_023617043.1 | XP_023472811.1 | 908 | 75 | |||
| XM_023617044.1 | XP_023472812.1 | 481 | 75 | |||
| XM_023617045.1 | XP_023472813.1 | 755 | 75 | |||
| XM_023617046.1 | XP_023472814.1 | 899 | 75 | |||
| XM_023617047.1 | XP_023472815.1 | 472 | 75 | |||
| XM_023617048.1 | XP_023472816.1 | 446 | 75 | |||
Set Membership: |
|