Search
Search

SH3TC2| Gene Symbol: SH3TC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005599310.3 | XP_005599367.1 | 3501 | 96 | ||
| XM_023617464.1 | XP_023473232.1 | 3505 | 96 | |||
| XM_023617466.1 | XP_023473234.1 | 3489 | 96 | |||
| XM_023617467.1 | XP_023473235.1 | 3171 | 96 | |||
| XM_023617468.1 | XP_023473236.1 | 3089 | 96 | |||