Search
Search

SLC23A1| Gene Symbol: SLC23A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014730641.2 | XP_014586127.1 | 2101 | 62 | ||
| XM_023617598.1 | XP_023473366.1 | 2102 | 62 | |||
| XM_023617599.1 | XP_023473367.1 | 924 | 62 | |||
| XM_023617600.1 | XP_023473368.1 | 1894 | 62 | |||
Set Membership: |
|