Search
Search

DNASE1L1| Gene Symbol: DNASE1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014729301.2 | XP_014584787.1 | 336 | 67 | ||
| XM_014729303.2 | XP_014584789.1 | 336 | 67 | |||
| XM_023633972.1 | XP_023489740.1 | 501 | 67 | |||
| XM_023633973.1 | XP_023489741.1 | 336 | 67 | |||
Set Membership: |
|