Search
Search
| Gene Symbol: VILL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014731784.2 | XP_014587270.1 | 1756 | 67 | ||
| XM_014731785.2 | XP_014587271.1 | 4425 | 67 | |||
| XM_014731786.2 | XP_014587272.1 | 4530 | 67 | |||
| XM_023620738.1 | XP_023476506.1 | 1688 | 67 | |||
| XM_023620739.1 | XP_023476507.1 | 4642 | 67 | |||
Set Membership: |
|