Search
Search

HTATSF1| Gene Symbol: HTATSF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001233031.5 | XP_001233032.1 | 8 - 9 | 1192 | 66 | |
| GenBank mRNA | AJ720151.1 | - | 8 - 9 | 1278 | 66 | |
| BX931656.1 | - | 7 - 8 | 786 | 66 | ||
Set Membership: |
|