Search
Search

BIRC6| Gene Symbol: BIRC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015283759.2 | XP_015139245.1 | 61 - 62 | 12451 | 72 | |
| XM_015283760.2 | XP_015139246.1 | 61 - 62 | 12439 | 72 | ||
| XM_015283761.2 | XP_015139247.1 | 61 - 62 | 12392 | 72 | ||
| XM_015283762.2 | XP_015139248.1 | 61 - 62 | 12352 | 72 | ||
| XM_015283763.2 | XP_015139249.1 | 61 - 62 | 12340 | 72 | ||
| XM_015283765.2 | XP_015139251.1 | 60 - 61 | 12304 | 72 | ||
| XM_015283766.2 | XP_015139252.1 | 61 - 62 | 12424 | 72 | ||
| XM_025149297.1 | XP_025005065.1 | 61 - 62 | 12461 | 72 | ||
| XM_419512.6 | XP_419512.5 | 61 - 62 | 12436 | 72 | ||
| GenBank mRNA | CR338711.2 | - | 8 - 9 | 1801 | 72 | |
Set Membership: |
|