Search
Search

EML5| Gene Symbol: EML5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015287733.2 | XP_015143219.1 | 12 - 13 | 2073 | 125 | |
| XM_025151282.1 | XP_025007050.1 | 12 - 13 | 2074 | 125 | ||
| XM_025151283.1 | XP_025007051.1 | 12 - 13 | 1924 | 125 | ||
| XM_025151284.1 | XP_025007052.1 | 12 - 13 | 1924 | 125 | ||
| XM_025151285.1 | XP_025007053.1 | 12 - 13 | 2073 | 125 | ||
| XM_421310.5 | XP_421310.2 | 12 - 13 | 2073 | 125 | ||
| GenBank mRNA | CR391271.1 | - | 4 - 5 | 584 | 125 | |
Set Membership: |
|