Search
Search

PHF6| Gene Symbol: PHF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004940662.2 | XP_004940719.1 | 3 - 4 | 661 | 118 | |
| XM_004940663.2 | XP_004940720.1 | 3 - 4 | 661 | 118 | ||
| XM_025149801.1 | XP_025005569.1 | 2 - 3 | 278 | 118 | ||
| XM_420223.5 | XP_420223.3 | 3 - 4 | 661 | 118 | ||
| GenBank mRNA | BX933944.1 | - | 3 - 4 | 663 | 118 | |
Set Membership: |
|