Search
Search

DUS1L| Gene Symbol: DUS1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001232802.5 | XP_001232803.2 | 7 - 8 | 953 | 58 | |
| XM_004946151.3 | XP_004946208.1 | 7 - 8 | 969 | 58 | ||
| GenBank mRNA | CR353630.1 | - | 7 - 8 | 899 | 58 | |
| CR406111.1 | - | 1 - 2 | 75 | 58 | ||
Set Membership: |
|