Search
Search

HNRNPLL| Gene Symbol: HNRNPLL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001305651.2 | NP_001292580.2 | 2 - 3 | 427 | 122 | |
| XM_015283458.2 | XP_015138944.1 | 2 - 3 | 377 | 122 | ||
| GenBank mRNA | BX930400.2 | - | 2 - 3 | 163 | 122 | |
| BX933171.1 | - | 2 - 3 | 427 | 122 | ||
| CR388661.1 | - | 2 - 3 | 391 | 122 | ||
Set Membership: |
|