Search
Search

RPGRIP1L| Gene Symbol: RPGRIP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001234247.5 | XP_001234248.3 | 2 - 3 | 544 | 79 | |
| XM_004944174.3 | XP_004944231.2 | 2 - 3 | 544 | 79 | ||
| XM_015292373.2 | XP_015147859.1 | 2 - 3 | 544 | 79 | ||
| XM_015292374.2 | XP_015147860.1 | 2 - 3 | 544 | 79 | ||
| XM_015292375.2 | XP_015147861.1 | 2 - 3 | 544 | 79 | ||
| XM_025154313.1 | XP_025010081.1 | 2 - 3 | 544 | 79 | ||
| GenBank mRNA | BX932540.1 | - | 2 - 3 | 431 | 79 | |
| BX934403.2 | - | 2 - 3 | 480 | 79 | ||
| BX934423.2 | - | 2 - 3 | 356 | 79 | ||
Set Membership: |
|