Search
Search

RECQL5| Gene Symbol: RECQL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204102.2 | NP_989433.2 | 4 - 5 | 906 | 75 | |
| XM_015295117.2 | XP_015150603.1 | 4 - 5 | 838 | 75 | ||
| XM_025141537.1 | XP_024997305.1 | 4 - 5 | 855 | 75 | ||
| GenBank mRNA | AB036057.1 | - | 4 - 5 | 862 | 75 | |
| AB074262.1 | - | 4 - 5 | 911 | 75 | ||
Set Membership: |
|