Search
Search

LRRC49| Gene Symbol: LRRC49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004943763.3 | XP_004943820.1 | 4 - 5 | 481 | 67 | |
| XM_004943764.3 | XP_004943821.1 | 3 - 4 | 425 | 67 | ||
| GenBank mRNA | BX931156.1 | - | 1 - 2 | 87 | 67 | |
Set Membership: |
|