Search
Search

KIZ| Gene Symbol: KIZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006394.1 | NP_001006394.1 | 4 - 5 | 457 | 61 | |
| XM_015283363.2 | XP_015138849.1 | 4 - 5 | 460 | 61 | ||
| XM_015283364.2 | XP_015138850.1 | 4 - 5 | 460 | 61 | ||
| XM_015283365.2 | XP_015138851.1 | 4 - 5 | 460 | 61 | ||
| XM_015283366.2 | XP_015138852.1 | 4 - 5 | 460 | 61 | ||
| XM_015283367.2 | XP_015138853.1 | 4 - 5 | 2413 | 61 | ||
| XM_015283369.2 | XP_015138855.1 | 4 - 5 | 2894 | 61 | ||
| XM_015283370.2 | XP_015138856.1 | 5 - 6 | 1220 | 61 | ||
| XM_015283371.2 | XP_015138857.1 | 4 - 5 | 1123 | 61 | ||
| XM_015283373.2 | XP_015138859.1 | 2 - 3 | 140 | 61 | ||
| XM_025148639.1 | XP_025004407.1 | 4 - 5 | 460 | 61 | ||
| XM_025148640.1 | XP_025004408.1 | 5 - 6 | 1281 | 61 | ||
| GenBank mRNA | AJ720271.1 | - | 4 - 5 | 457 | 61 | |
Set Membership: |
|