Search
Search

GATAD2A| Gene Symbol: GATAD2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012552.2 | NP_001012570.2 | 6 - 7 | 911 | 59 | |
| XM_015300041.2 | XP_015155527.2 | 8 - 9 | 3462 | 59 | ||
| XM_015300042.2 | XP_015155528.1 | 8 - 9 | 1125 | 59 | ||
| XM_015300043.2 | XP_015155529.1 | 8 - 9 | 1139 | 59 | ||
| XM_015300044.2 | XP_015155530.1 | 8 - 9 | 1125 | 59 | ||
| XM_015300045.2 | XP_015155531.1 | 7 - 8 | 947 | 59 | ||
| XM_015300046.2 | XP_015155532.1 | 7 - 8 | 1034 | 59 | ||
| XM_025144269.1 | XP_025000037.1 | 8 - 9 | 3462 | 59 | ||
| XM_025144270.1 | XP_025000038.1 | 8 - 9 | 3471 | 59 | ||
| XM_025144271.1 | XP_025000039.1 | 7 - 8 | 3372 | 59 | ||
| XM_025144272.1 | XP_025000040.1 | 7 - 8 | 948 | 59 | ||
| XM_025144273.1 | XP_025000041.1 | 7 - 8 | 1049 | 59 | ||
| GenBank mRNA | AJ851822.1 | - | 7 - 8 | 965 | 59 | |
| CR391510.1 | - | 2 - 3 | 283 | 59 | ||
Set Membership: |
|