Search
Search

KLHL14| Gene Symbol: KLHL14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012866.1 | NP_001012884.1 | 3 - 4 | 1310 | 103 | |
| GenBank mRNA | AJ851615.1 | - | 3 - 4 | 1310 | 103 | |
| CR387415.1 | - | 2 - 3 | 103 | |||
Set Membership: |
|