Search
Search

ACTN2| Gene Symbol: ACTN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205323.1 | NP_990654.1 | 17 - 18 | 2440 | 121 | |
| GenBank mRNA | X13874.1 | - | 2207 | 121 | ||
| X59247.1 | - | 2440 | 121 | |||
Set Membership: |
|