Search
Search

INCENP| Gene Symbol: INCENP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205330.1 | NP_990661.1 | 15 - 16 | 2342 | 65 | |
| XM_015286759.2 | XP_015142245.1 | 16 - 17 | 2483 | 65 | ||
| XM_015286760.2 | XP_015142246.1 | 15 - 16 | 2371 | 65 | ||
| GenBank mRNA | Z25419.1 | - | 2342 | 65 | ||
| Z25420.1 | - | 2456 | 65 | |||
Set Membership: |
|