Search
Search

VPS33B| Gene Symbol: VPS33B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031348.1 | NP_001026519.1 | 14 - 15 | 1160 | 67 | |
| XM_015292119.2 | XP_015147605.1 | 14 - 15 | 1146 | 67 | ||
| GenBank mRNA | AJ719863.1 | - | 14 - 15 | 1160 | 67 | |
| CR354273.1 | - | 5 - 6 | 353 | 67 | ||
| CR385810.1 | - | 14 - 15 | 1131 | 67 | ||
Set Membership: |
|