Search
Search

VPS33B| Gene Symbol: VPS33B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031348.1 | NP_001026519.1 | 18 - 19 | 1458 | 71 | |
| XM_015292119.2 | XP_015147605.1 | 18 - 19 | 1444 | 71 | ||
| GenBank mRNA | AJ719863.1 | - | 18 - 19 | 1458 | 71 | |
| CR354273.1 | - | 9 - 10 | 651 | 71 | ||
| CR385810.1 | - | 18 - 19 | 1429 | 71 | ||
Set Membership: |
|