Search
Search

ST7| Gene Symbol: ST7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105672.1 | NP_001099142.1 | 7 - 8 | 793 | 81 | |
| XM_015281094.2 | XP_015136580.1 | 7 - 8 | 1030 | 81 | ||
| XM_015281100.2 | XP_015136586.1 | 7 - 8 | 1144 | 81 | ||
| XM_025147633.1 | XP_025003401.1 | 7 - 8 | 802 | 81 | ||
| XM_025147634.1 | XP_025003402.1 | 7 - 8 | 831 | 81 | ||
| XM_025147642.1 | XP_025003410.1 | 7 - 8 | 664 | 81 | ||
| GenBank mRNA | BX934627.2 | - | 7 - 8 | 1002 | 81 | |
Set Membership: |
|