Search
Search

SLC25A3| Gene Symbol: SLC25A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006236.1 | NP_001006236.1 | 8 - 9 | 910 | 76 | |
| XM_015284675.2 | XP_015140161.1 | 7 - 8 | 1012 | 76 | ||
| XM_015284680.2 | XP_015140166.1 | 7 - 8 | 1084 | 76 | ||
| XM_025148247.1 | XP_025004015.1 | 6 - 7 | 926 | 76 | ||
| GenBank mRNA | AJ719594.1 | - | 8 - 9 | 910 | 76 | |
Set Membership: |
|