Search
Search

APLP2| Gene Symbol: APLP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006317.3 | NP_001006317.2 | 17 | 2118 | 65 | |
| GenBank mRNA | AJ719398.1 | - | 4 | 354 | 65 | |
| CO422404.1 | - | 3 | 206 | 65 | ||
Set Membership: |
|