Search
Search

SLC35G2| Gene Symbol: SLC35G2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031299.1 | NP_001026470.1 | 2 - 3 | 194 | 72 | |
| XM_015291338.2 | XP_015146824.1 | 2 - 3 | 280 | 72 | ||
| XM_015291339.2 | XP_015146825.1 | 2 - 3 | 366 | 72 | ||
| XM_015291340.2 | XP_015146826.1 | 2 - 3 | 202 | 72 | ||
| GenBank mRNA | AJ720290.1 | - | 2 - 3 | 194 | 72 | |
Set Membership: |
|