Search
Search

DNAJC6| Gene Symbol: DNAJC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177415.2 | NP_001170886.1 | 8 - 9 | 1010 | 58 | |
| XM_015291026.2 | XP_015146512.1 | 9 - 10 | 1208 | 58 | ||
| XM_015291027.2 | XP_015146513.1 | 9 - 10 | 1397 | 58 | ||
Set Membership: |
|