Search
Search

ARIH2| Gene Symbol: ARIH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199221.1 | NP_001186150.1 | 14 - 15 | 1556 | 63 | |
| XM_015293105.2 | XP_015148591.1 | 15 - 16 | 1718 | 63 | ||
| XM_025154483.1 | XP_025010251.1 | 15 - 16 | 1668 | 63 | ||
| GenBank mRNA | MK335707.1 | - | 13 - 14 | 1394 | 63 | |
| MK335708.1 | - | 13 - 14 | 1394 | 63 | ||
| MK335709.1 | - | 13 - 14 | 1394 | 63 | ||
Set Membership: |
|