Search
Search

BRSK1| Gene Symbol: BRSK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199596.1 | NP_001186525.1 | 7 - 8 | 617 | 67 | |
| XM_015286601.2 | XP_015142087.1 | 7 - 8 | 1020 | 67 | ||
| XM_015286602.2 | XP_015142088.1 | 7 - 8 | 1020 | 67 | ||
| XM_015286603.2 | XP_015142089.1 | 7 - 8 | 1020 | 67 | ||
| XM_015286604.2 | XP_015142090.1 | 7 - 8 | 1020 | 67 | ||
| XM_015286605.2 | XP_015142091.1 | 7 - 8 | 1020 | 67 | ||
| XM_015286606.2 | XP_015142092.1 | 7 - 8 | 1020 | 67 | ||
| XM_015286608.2 | XP_015142094.1 | 7 - 8 | 637 | 67 | ||
| XM_015286609.2 | XP_015142095.1 | 7 - 8 | 637 | 67 | ||
| XM_015286610.2 | XP_015142096.1 | 7 - 8 | 1020 | 67 | ||
| XM_015286611.2 | XP_015142097.1 | 7 - 8 | 1019 | 67 | ||
| GenBank mRNA | BU392070.1 | - | 4 - 5 | 311 | 67 | |
Set Membership: |
|